Human Recombinant ACYP2 (from E. coli), GST Tag
Supplier: Sino Biological
10437-H09E-20
CA76454-762
Human Recombinant ACYP2 (from E. coli), GST Tag
Proteins and Peptides
A DNA sequence encoding the human ACYP2 (P14621) (Ser 2-Tyr 99) was fused with the GST tag at the N-terminus.