Human Recombinant VE-Cadherin/CD144/CDH5 Protein (His & Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10433-H03H-50 10433-H03H-100 10433-H03H-50
103621-212EA 300.98 USD
CA76454-758 103621-212 103679-226
Human Recombinant VE-Cadherin/CD144/CDH5 Protein (His & Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human CDH5 (NP_001786.2) (Met 1-Gln 593) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.