Human Recombinant TROP2/TACSTD2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10428-H08H-1 10428-H08H-100
76642-172EA 1156 USD
76642-172 103621-202
Human Recombinant TROP2/TACSTD2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Thr274) of human TROP2 (NP_002344.2) was expressed fused with a polyhistidine tag at the C-terminus.