Human/Mouse/Rat/Rhesus/Canine Recombinant BMP-2/BMP2A Protein (from E. coli)
Supplier: Sino Biological
10426-HNAE-5 10426-HNAE-5 10426-HNAE-1 10426-HNAE-20 10426-HNAE-1 10426-HNAE-20 10426-HNAE-100
103624-066EA 382.48 USD
CA76453-598 103624-066 CA76453-596 CA76454-746 103677-568 103677-566 103679-214
Human/Mouse/Rat/Rhesus/Canine Recombinant BMP-2/BMP2A Protein (from E. coli)
Proteins and Peptides
A DNA sequence encoding the mature form of human / mouse / rat / rhesus / canine BMP2 (NP_001191.1) (Gln283-Arg396) was expressed with an initial Met. The mature form sequences of human, mouse, rat, rhesus and canine BMP2 are identical.