Human Recombinant TNFRSF1B (from HEK293 Cells), His Tag
Supplier: Sino Biological
10417-H08H-100 10417-H08H-50 10417-H08H-50 10417-H08H-1
103679-198EA 178 USD
103679-198 CA76454-730 103621-186 76641-908
Human Recombinant TNFRSF1B (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Asp 257) of human TNFRII (NP_001057.1) was expressed, fused with a C-terminal polyhistidine tag.