Human Recombinant KLK7 (from HEK293 cells), His Tag
Supplier: Sino Biological
10416-H08H-10 10416-H08H-20 10416-H08H-10 10416-H08H-20
103679-194EA 320.35 USD
103679-194 103679-192 CA76454-724 CA76454-726
Human Recombinant KLK7 (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human KLK7 (NP_005037.1) extracellular domain (Met 1-Arg 253) was expressed, with a C-terminal polyhistidine tag.