Human Recombinant CD4/LEU3 Protein (aa 1-208, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10400-H08H1-100
103632-612EA 300.98 USD
103632-612
Human Recombinant CD4/LEU3 Protein (aa 1-208, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human CD4 (NP_000607.1) N-terminal two domains (D1D2) (Met 1-Ser 208) was fused with a polyhistidine tag at the C-terminus.