Human Recombinant IL20RA (from HEK293 cells)
Supplier: Sino Biological
10397-H08H-50
CA76454-698
Human Recombinant IL20RA (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain of human IL20Rα isoform 1 (NP_055247.21) (Met 1-Lys 250) was fused with a polyhistidine tag at the C-terminus.