Human Recombinant HAVCR2 (from HEK293 cells)
Supplier: Sino Biological
10390-H03H-50 10390-H03H-100 10390-H03H-50
103679-144EA 178 USD
CA76454-676 103679-144 103621-130
Human Recombinant HAVCR2 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the human TIMD3 (NP_116171.3) extracellular domain (Met 1-Arg 200) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.