Human Recombinant TNFSF14 (from HEK293 Cells), His Tag
10386-H07H-5 10386-H07H-20
CA76455-284 CA76453-552
Human Recombinant TNFSF14 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human TNFSF14 (NP_003798.2) extracellular domain (Asp 74-Val 240) was fused with a polyhistidine-tag at the N-terminus.