Human Recombinant Thrombopoietin/THPO/TPO Protein (His Tag) (from CHO cells)
Supplier: Sino Biological
10381-H08C-20 10381-H08C-5 10381-H08C-1 10381-H08C-20 10381-H08C-5 10381-H08C-100
103677-508EA 83.85 USD
103677-508 76778-362 103621-094 103679-136 CA76454-668 CA76453-548
Human Recombinant Thrombopoietin/THPO/TPO Protein (His Tag) (from CHO cells)
Proteins and Peptides
A DNA sequence encoding the human THPO (NP_000451.1) precursor (Met 1-Gly 353) was fused with a polyhistidine tag at the C-terminus.