Human Recombinant THPO (from HEK293 Cells), His Tag
Supplier: Sino Biological
10381-H07H-20 10381-H07H-20 10381-H07H-5 10381-H07H-1 10381-H07H-100 10381-H07H-5
76641-086EA 2258 USD
76641-086 CA76453-546 103621-092 CA76454-666 103677-506 103679-134
Human Recombinant THPO (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the mature form of human THPO (NP_000451.1) (Ser 22-Gly 353) was fused with a polyhistidine tag at the N-terminus.