Human Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10377-H02H-50 10377-H02H-50 10377-H02H-500
103679-122EA 179.78 USD
CA76454-654 103679-122 76641-038
Human Recombinant PD1/PDCD1/CD279 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human PDCD1 (NP_005009.2) (Met1-Gln167) was expressed with the Fc region of human IgG1 at the C-terminus.