Human Recombinant CD32a/FCGR2A Protein (167 His, His & AVI Tag), Biotinylated (from HEK293 Cells)
Supplier: Sino Biological
10374-H27H1-B-20 10374-H27H1-B-100
76642-416EA 688 USD
76642-416 103633-780
Human Recombinant CD32a/FCGR2A Protein (167 His, His & AVI Tag), Biotinylated (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met1-Met210) of human CD32a (P12318-1) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.