Human Recombinant FCGR2A (from HEK293 cells)
Supplier: Sino Biological
10374-H27H-50 10374-H27H-100 10374-H27H-50
103679-120EA 178 USD
103679-120 103621-080 CA76454-652
Human Recombinant FCGR2A (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Met 210) of human CD32a (AAA35827.1) was fused with a C-terminal polyhistidine tagged AVI tag at the C-terminus. It is identical to FCGR2A131H/R in the reference.