Human Recombinant FCGR2A (from CHO cells), His Tag
Supplier: Sino Biological
10374-H08C-50 10374-H08C-50
103679-116EA 178 USD
CA76454-648 103679-116
Human Recombinant FCGR2A (from CHO cells), His Tag
Proteins and Peptides
A DNA sequence encoding (Met 1-Ile 218) of human CD32a (AAA35827.1) was expressed with a C-terminal polyhistidine tag. It is identical to FCGR2A131H/R in the reference.