Human Recombinant FRZB (from HEK293 cells), His Tag
Supplier: Sino Biological
10370-H08H-20
CA76454-634
Human Recombinant FRZB (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human FRZB (NP_001454.2) (Ala32-Asn325) was expressed with a C-terminal polyhistidine tag.