Human Recombinant ERBB4 (from HEK293 cells), His Tag
Supplier: Sino Biological
10363-H08H-50 10363-H08H-50
103679-088EA 178 USD
103679-088 CA76454-620
Human Recombinant ERBB4 (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human ERBB4 (NP_005226.1) (Met1-Arg649) with a C-terminal polyhistidine tag was expressed. Human and Rhesus ERBB4 sequences are identical.