Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ErbB4 Protein, Human, Recombinant

Supplier: Sino Biological
10363-H02H-100 10363-H02H-50
A DNA sequence encoding the human ERBB4 (NP_005226.1) (Met1-Arg649) was expressed, fused with the Fc region of human IgG1 at the C-terminus.Human and Rhesus ERBB4 sequences are identical.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µg 10363-H02H-100 103621-058 Each (100µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

50 µg 10363-H02H-50 103679-082 Each (50µG) Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name ERBB4
Protein/Peptide Type Recombinant
Species Human
Source HEK293 cells
Gene ID NP_005226.1
Sequence Met1-Arg649
Purity > 85 % as determined by SDS-PAGE
Endotoxin Content < 1.0 EU per μg of the protein as determined by the LAL method
Molecular Weight The recombinant human ERBB4/Fc is a disulfide-linked homodimer. The reduced monomer comprises 865 amino acids and has a predicted molecular mass of 96.6 kDa. The apparent molecular mass of the protein is approximately 117 kDa in SDS-PAGE under reducing conditions.
Formulation Lyophilized from sterile PBS, pH7.4.
Biological Activity 1. Measured by its ability to bind biotinylated human Fc-NRG1 (isoform Beta1) (Cat:11609-H01H) in a functional ELISA. 2. Measured by its ability to bind biotinylated human NRG1 (isoform Beta1) (Cat:11609-H01H2) in a functional ELISA. 3. Measured by its ability to bind biotinylated human NRG1 (aa 2-246)-Fc (Cat:11609-H04H) in a functional ELISA.
Reconstitution Instructions A hardcopy of COA with reconstitution instruction is sent along with the products. Please refer to it for detailed information.
Storage Conditions Store it under sterile conditions at -20℃ to -80℃. It is recommended that the protein be aliquoted for optimal storage. Avoid repeated freeze-thaw cycles.
Shipping Temperature In general, recombinant proteins are provided as lyophilized powder which are shipped at ambient temperature.<br>Bulk packages of recombinant proteins are provided as frozen liquid. They are shipped out with blue ice unless customers require otherwise.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty