Human Recombinant IL-15/IL15/Interleukin 15 Protein (His Tag) (from E. coli)
Supplier: Sino Biological
10360-H07E-5 10360-H07E-10 10360-H07E-5 10360-H07E-10 10360-H07E-100
103677-496EA 62.35 USD
103677-496 103679-076 CA76453-536 103621-056 CA76454-608
Human Recombinant IL-15/IL15/Interleukin 15 Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human IL15 (NP_000576.1) (Asn49-Ser162) was expressed with a polyhistidine tag at the N-terminus.