Human Recombinant RBP4 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10354-H08H-50 10354-H08H-100 10354-H08H-50
103621-038EA 300.98 USD
103621-038 103679-062 CA76454-594
Human Recombinant RBP4 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the pro form of human RBP4 (NP_006735.2) (Met 1-Leu 201) was expressed with a C-terminal polyhistidine tag.