Human Recombinant IFNW1 (from HEK293 cells), His Tag
Supplier: Sino Biological
10353-H08H-20
CA76454-592
Human Recombinant IFNW1 (from HEK293 cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human IFNW1 (P05000) (Met1-Ser195) was expressed with a polyhistidine tag at the C-terminus.