Human Recombinant IFNA10 (from HEK293 cells)
Supplier: Sino Biological
10349-H01H-5 10349-H01H-50
CA76453-532 CA76454-580
Human Recombinant IFNA10 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the human IFNA10 (P01566) (Cys 24-Asp 189) was fused with the Fc region of human IgG1 at the N-terminus.