Human Recombinant ICAM1 (from HEK293 cells)
Supplier: Sino Biological
10346-HCCH-50
CA76454-570
Human Recombinant ICAM1 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the human ICAM1 (NP_000192.2) extracellular domain (Met 1-Glu 480) with five amino acids (DDDDK) at the C-terminus was expressed and purified.