Human Recombinant ICAM1 (from HEK293 cells)
Supplier: Sino Biological
10346-H03H-100 10346-H03H-50 10346-H03H-50
103679-032EA 116.1 USD
103679-032 CA76454-564 103622-440
Human Recombinant ICAM1 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the human ICAM1 (NP_000192.2) extracellular domain (Met 1-Glu 480) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.