Human Recombinant SLITRK1 (from HEK293 Cells)
Supplier: Sino Biological
10340-H03H-50
CA76454-818
Human Recombinant SLITRK1 (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Ser 616) of human SLITRK1 (NP_443142.1) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.