Human Recombinant SELE (from HEK293 Cells)
Supplier: Sino Biological
10335-H03H-200 10335-H03H-100
103622-424EA 212.85 USD
103622-424 103622-426
Human Recombinant SELE (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Pro 556) of human CD62E (NP_000441.2) precursor was expressed with the fused C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.