Human Recombinant ICAM3 (from HEK293 cells)
Supplier: Sino Biological
10333-H03H-50
CA76454-544
Human Recombinant ICAM3 (from HEK293 cells)
Proteins and Peptides
The extracellular domain (Met 1-His 485) of human ICAM3 (NP_002153.2) precursor was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.