Human Recombinant ICAM-2/CD102 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10332-H08H-50 10332-H08H-50 10332-H08H-100
103679-010EA 212.85 USD
103679-010 CA76454-542 103620-994
Human Recombinant ICAM-2/CD102 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Gln 223) of human ICAM2 (NP_000864.2) was fused with a polyhistidine tag at the C-terminus.