Human Recombinant FLT3LG (from Baculovirus (Insect cells)), His Tag
Supplier: Sino Biological
10315-H07B-20
CA76454-422
Human Recombinant FLT3LG (from Baculovirus (Insect cells)), His Tag
Proteins and Peptides
A DNA sequence encoding the human FLT3LG (P49771-1) (Thr27-Pro185) was expressed, with a polyhistidine tag at the N-terminus.