Human Recombinant POSTN (from HEK293 Cells), His Tag
10299-H08H-50
CA76454-396
Human Recombinant POSTN (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human POSTN isoform 3 (NP_001129407.1) corresponding to amino acid (Met 1-Gln 781) was expressed with a C-terminal polyhistidine tag.