Human Recombinant SerpinE1 (from HEK293 Cells), His Tag
Supplier: Sino Biological
10296-H08H-1 10296-H08H-10 10296-H08H-50 10296-H08H-20 10296-H08H-10 10296-H08H-20
76640-910EA 2268 USD
CA76454-386 CA76454-388 76640-910 103678-838 76641-988 103678-840
Human Recombinant SerpinE1 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human SerpinE1 precursor (NP_000593.1) (Met 1-Pro 402) was expressed with a C-terminal polyhistidine tag.