Human Recombinant SerpinD1 (from HEK293 Cells), His Tag
Supplier: Sino Biological
10295-H08H-10 10295-H08H-50 10295-H08H-20 10295-H08H-10 10295-H08H-20
76778-684EA 311.41 USD
76778-684 CA76454-384 103678-836 103678-834 CA76454-382
Human Recombinant SerpinD1 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the human SerpinD1 precursor (NP_000176.2) (Met 1-Ser 499) was expressed with a C-terminal polyhistidine tag.