Human Recombinant PDCD1LG2 (from HEK293 Cells), His Tag
Supplier: Sino Biological
10292-H08H-200 10292-H08H-1 10292-H08H-100
103622-390EA 351.48 USD
103622-390 103622-392 76641-442
Human Recombinant PDCD1LG2 (from HEK293 Cells), His Tag
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Pro 219) of human PD-L2 (NP_079515.2) was expressed with a C-terminal polyhistidine tag.