Human Recombinant NTF3 (from E. coli)
Supplier: Sino Biological
10286-HNAE-5 10286-HNAE-10
CA76453-518 CA76454-360
Human Recombinant NTF3 (from E. coli)
Proteins and Peptides
A DNA sequence encoding the human NT3 (P20783-1) (Tyr139-Thr257) was expressed and purified.