Human Recombinant MMP7 (from HEK293 Cells)
Supplier: Sino Biological
10277-H01H-20 10277-H01H-20 10277-H01H-10 10277-H01H-10
103679-268EA 298 USD
CA76454-798 103679-268 CA76454-800 103679-266
Human Recombinant MMP7 (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the active form of human MMP7 (NP_002414.1) (Tyr 95-Lys 267) was expressed with the fused Fc region of human IgG1 at the N-terminus.