Human Recombinant PDGFC (from Yeast), His Tag
Supplier: Sino Biological
10273-H07Y-100 10273-H07Y-20 10273-H07Y-20
103620-940EA 481.6 USD
103620-940 CA76454-316 103678-742
Human Recombinant PDGFC (from Yeast), His Tag
Proteins and Peptides
A DNA sequence encoding the human PDGFC (NP_057289.1) (Val235-Gly345) was expressed with a polyhistidine tag at the N-terminus.