Human Recombinant OMGP/OMG Protein (aa 1-420, His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10269-H08H1-20 10269-H08H1-50
103622-368EA 148.35 USD
103622-368 103622-370
Human Recombinant OMGP/OMG Protein (aa 1-420, His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human OMGP (P23515-1) (Met 1-Asn 420) without the pro peptide was expressed, fused with a polyhistidine tag at the C-terminus.