Human Recombinant FGF9 Protein (Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10262-H01H-20 10262-H01H-100 10262-H01H-20 10262-H01H-5 10262-H01H-5
103678-890EA 139.38 USD
103678-890 103622-348 CA76454-438 103677-448 CA76453-504
Human Recombinant FGF9 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the mature form of human fibroblast growth factor 9 (NP_002001.1) (Leu 4-Ser 208) was expressed with the fused Fc region of human IgG1 at the N-terminus.