CK-666

Synonyms: 2-Fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]-benzamide

B2147-25 B2147-5
76290-094EA 343.88 USD
76290-094 76290-092
CK-666
CK-666

A potent, selective and reversible inhibitor of ARP2/3 complex


CK-666 is a potent, selective and reversible inhibitor of ARP2/3 complex (actin related proteins 2 and 3) with an IC₅₀ = 4 µM for human ARP2/3. Blocks actin nucleation and radically alters lamellipodial actin architecture, suspended cell shape and the cell spreading process. CK-666 attenuates BBB dysfunction induced by methamphetamine. A key new tool for studying actin assembly and the diverse range of actin-related processes in normal and pathophysiology.

Formula: C₁₈H₁₇FN₂O
MW: 296.3 g/mol
Storage Temperature: Freezer
CAS Number: 442633-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR