Amprenavir

Synonyms: (S)-Tetrahydrofuran-3-yl ((2S,3R)-4-((4-amino-N-isobutylphenyl)sulphonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate(S)-Tetrahydrofuran-3-yl ((2S,3R)-4-((4-amino-N-isobutylphenyl)sulfonamido)-3-hydroxy-1-phenylbutan-2-yl)carbamate

B2141-25 B2141-5
76289-904EA 757.72 USD
76289-904 76289-822
Amprenavir
Amprenavir

A HIV Protease Inhibitor


Amprenavir is the active metabolite of the pro-drug fosamprenavir. It acts as an antiretroviral HIV Protease Inhibitor. When used alone or in combination therapy with other antiretroviral agents, amprenavir decreases viral load and increases CD4+ cell counts.

Formula: C₂₅H₃₅N₃O₆S
MW: 505.6269 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00934214
CAS Number: 161814-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR