Ritonavir

Synonyms: Thiazol-5-ylmethyl ((2S,3S,5S)-3-hydroxy-5-((S)-2-(3-((2-isopropylthiazol-4-yl)methyl)-3-methylureido)-3-methylbutanamido)-1,6-diphenylhexan-2-yl)carbamateAbbott 84538ABT-538A-84538

B2140-10 B2140-50
76289-818EA 92.82 USD
76289-818 76289-820
Ritonavir
Ritonavir

An orally bioavailable HIV-1 and HIV-2 protease inhibitor


Ritonavir is an orally bioavailable HIV-1 and HIV-2 protease inhibitor (EC₅₀ values are 0.022-0.13 and 0.16 μM, respectively). Through CYP3A4 inhibition, low doses of ritonavir can reduce the metabolism of concomitantly administered protease inhibitors, enhancing their bioavailability and efficacy.

Formula: C₃₇H₄₈N₆O₅S₂
MW: 720.94 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00927142
CAS Number: 155213-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR