Enrofloxacin

Danger

Synonyms: 1-Cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid

B2101-500 B2101-100
76290-042EA 81.43 USD
76290-042 76290-044
Enrofloxacin
Enrofloxacin

A broad-spectrum fluoroquinolone antibiotic


Enrofloxacin is a broad-spectrum fluoroquinolone antibiotic that inhibits bacterial DNA gyrase. Enrofloxacin displays high bioavailability in most species and rapid absorption after oral, intramuscular, or subcutaneous administration. Enrofloxacin demonstrates significant post-antibiotic effect for both Gram-negative and Gram-positive bacteria and is active in both stationary and growth phases of bacterial replication.

Formula: C₁₉H₂₂FN₃O₃
MW: 359.40 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00792463
CAS Number: 93106-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR