Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Dibismuth trioxide
, Bismuth Trioxide
Management of Change (MOC) category = RL
MACR1057-24
MACR1057-24
MACR1057-24
MK1057-24CS
17795.02
USD
MK1057-24
JT1057-24
CAMK105724
Bismuth(III) oxide ≥99.7%, AR®, Macron Fine Chemicals™
Bismuth (III) oxide
Formula:
Bi₂O₃ MW: 465.96 g/mol Density: 8.90 g/cm³ |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 |
Specification Test Results
CAUTION: For Manufacturing, processing or repackaging | |
Assay (Bi₂O₃) | ≥ 99.7 % |
pH (by aqueous extraction) | 6.75 - 9.25 |
Appearance (yellow powder) | Passes Test |
Arsenic (As) | ≤ 0.0008 % |
Chloride (Cl) | ≤ 0.001 % |
Copper (Cu) | ≤ 0.01 % |
Iron (Fe) | ≤ 0.01 % |
Sodium (Na) | ≤ 0.03 % |
On U.S. No. 325 Sieve | ≤ 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...