Drosophila Plug, Tradewinds
Supplier: TradeWinds (TWD)
51-17725
490016-764CS 403.08 USD
490016-764
Drosophila Plug, Tradewinds
Stoppers Cellulose Stoppers
Drosophila Plugs are high density foam--minimal shedding as well as reduced evaporation. Stretched yet maintains tight fit after repeated removal and closing.
  • Flat surface accepts marking pen
  • Superior barrier to mite infestation