Heidolph® MR Hei-Connect Magnetic Hotplate Stirrers
Supplier: Heidolph
Heidolph®
036110602 036110601 036110603 036110604
76291-930EA 2464.1 USD
76291-930 76291-940 76291-938 76291-936
Heidolph® MR Hei-Connect Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers
These hotplate stirrers feature excellent heat conduction and distribution as well as resistance against scratches and chemicals.
  • IP 32 protection glass
  • Program ramps and interval processes options are available
  • This unit features a digital display and allows for easy setting and monitoring of values in the illuminated readout
  • An illuminated button indicates clearly if the stirring mode is activated
Round hotplate is constructed of Silumin aluminum alloy (Kera-Disk®) with a ceramic coating. A separate on/off switch for heating prevents an unintentional heat-up. If heating is switched on the on/off button is illuminated for visual control. In addition, a residual heat indicator prevents from burning hazards when heat function is switched off.
For the protection an independent safety circuit will switch off heating if hotplate temperature exceeds 25 °C over set temperature.
Upgrade this magnetic stirrer with the optional PT1000 temperature sensor for precise temperature control, overshoot protection and reproducible results. MR Hei-Connect includes all features of the MR Hei-Tec unit but comes with an additional RS232 interface.
Delivery information: Supplied with Hei-Control software.