Would you like to log in again?
Total D-Amino Acid Assay Kit, BioVision, Inc.
Supplier: BioVision
K445-100
Total D-amino acid assay kit (Fluorometric) provides a quick, specific and easy method for measuring total D-Amino acid concentrations in a wide variety of samples.
  • Simple procedure, takes ~ 2 hours
  • Fast and convenient
  • The kit is simple to use, sensitive and high-throughput adaptable
  • Detection limit: 1.9 μM
D-Amino acids are present in both eukaryotes and prokaryotes. D-Amino acids act as a component of the cell wall in prokaryotes and a neurological regulator in eukaryotes. In mammals, D-Amino acids play an important role in physiological functions and their levels are regulated by D-Amino acid oxidase in the central nervous systems. Some important D-Amino acids include D-serine, D-aspartate and D-alanine. D-serine acts as a modulator of N-Methyl-D-aspartate receptor (NMDA receptor, NMDAR), while D-aspartate and D-alanine have been found to be elevated in the white and the gray matter respectively in patients’ brains suffering Alzheimer’s disease.
The abnormal levels of D-Amino acids could be used as a marker of neurological diseases: total D-Amino acid concentration in cerebrospinal fluid (CSF) has been shown to be higher in patients with Alzheimer’s disease when compared to samples from healthy donors.
In this assay, D-Amino acids are converted into an intermediate by the DAA enzyme mix that will further react with a probe to produce a strong fluorescence signal (Ex/Em= 535/587 nm).
ORDER
Expand All / Collapse All
Description No. of Assays/Kit Supplier No. VWR Catalog Number Unit Price Quantity
Total D-Amino Acid Assay Kit 100 K445-100 76289-458 Each Retrieving Restricted
Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty