Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AMV RTASE, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11EMAMV200 11EMAMV203
High efficiency enzyme from moloney murine leukemia virus.

  • From avian myeloblastosis virus
  • Withstands temperatures of 55 to 60 °C
  • RNase H and unwinding activity
  • Synthesis of full length copies of cDNA -10 kb
  • Low RNase H activity
  • Recombinant enzyme

AMV Reverse Transcriptase is an RNA-dependent DNA polymerase. The enzyme is composed of two polypeptides. The molecular weight
of the one subunit is 68000 Da and that of the other subunit 92000 Da. It is widely used for first and second strand synthesis of complementary DNA (cDNA) from mRNA templates for cloning or for hybridization probes. Such synthesis requires a primer, commonly oligo(dT) bound to the 3'-poly(A) region of mRNA. Polymerization proceeds in the 5' to 3' direction with no 5' to 3' exonuclease activity. Under the right conditions, high yields of full length cDNA can be obtained using AMV reverse transcriptase. AMV Reverse Transcriptase is suitable for synthesis of cDNA using specific primers, at temperatures of 55 to 60 °C. This high reaction temperature permits transcription of RNA containing secondary structure.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
200 U 11EMAMV200 IC11EMAMV200 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

3×200 U 11EMAMV203 IC11EMAMV203 Each Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Storage Conditions Store at –20 °C.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty