Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Secondary Pressure Vessels System, Saf-T-Pak

Supplier: SAF-T-PAK INC MS
STP-710 STP-701 STP-714 STP-740 STP-730 STP700
Secondary pressure vessels are pressure tested to hold an internal pressure differential of 95 kPa without leaking. The product is a two-part system consisting of an inner outer envelope they are stronger than today’s present technology.

  • Biohazard and leakproof poly bag
  • Outer envelope container of Tyvek
  • Each unit is for single use and is disposable

Infectious shipments: This system is the tested part of the shipperand restricted to that shipper.

Diagnostic shipments: this system is readily available for any specimen shipment using any of the versatile secondary pressure vessel system.



 

ORDER

Expand All / Collapse All
Description Dimensions Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Secondary Pressure Vessel System, Small Size Specimen 33/4×61/2" 50/Case STP700 MSPP-STP700 Case of 50 Retrieving
Secondary Pressure Vessel System, Small Size Specimen 33/4×61/2" 250/Case STP-701 MSPP-STP701 Case of 250 Retrieving
Secondary Pressure Vessel System, Versatile 71/2×91/2" 50/Case STP-710 MSPP-STP-710 Case of 50 Retrieving
Secondary Pressure Vessel System, Patented for DRY ICE 71/2×101/2" 50/Case STP-714 MSPP-STP714 Case of 50 Retrieving

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Secondary Pressure Vessel System, Medium Grid Box 10×10" 50/Case STP-730 MSPP-STP-730 Case of 50 Retrieving
Secondary Pressure Vessel System, Large Size for Transport of Forensic 12×16" 50/Case STP-740 MSPP-STP-740 Case of 50 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty