Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ISIS DNA Polymerase, MP Biomedicals

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11EPSIS100 11EPSIS103
For high fidelity PCR.

  • Extremely low error rates
  • Optimal blunt end cloning, Storage: –20 °C
  • The most thermostable polymerase available
  • Storage buffer: Tris HCl 20 mM, pH 8.0, KCl 100 mM, EDTA 0.1 mM, Dithiothreitol 1 mM, Glycerol 50%
  • Incubation: TrisHCl 20 mM, pH (25 °C) 9.0, KCl 25 mM, (NH₄)2SO₄
  • buffer 1X: MgSO₄ 1.5 mM, BSA 0.1 mg/ml Incubation buffer is provided 10X
  • Quality control: Activity, absence of nickases and endonucleases, absence of ribonucleases,specific PCR on genomic and phage
  • templates
  • Unit defi nition: One unit is the amount of enzyme required to catalyze the incorporation of 10 nmol of nucleosides into acid insoluble
  • material in 30 min. at 74 °C under assay conditions

Isis DNA Polymerase does not require primer 3'-end protection, unlike many other proofreading enzymes. This saves time during reaction setup and optimization. Isis DNA polymerase is able to amplify up to 6 kb on genomic DNA templates and 18 kb on phage DNA under a variety of amplification conditions. Isis DNA polymerase offers robust activity and fl exibility. There is a patent pending on the enzyme Isis DNA polymerase. Isis DNA Polymerase has been developed in collaboration with IFREMER.

Application includes: Nucleic Acid Amplification; PCR; High Fidelity PCR; Proofreading PCR.




 

ORDER

Expand All / Collapse All
Description Size Supplier No. VWR Catalog Number Unit Price Quantity
Proteins, Enzymes and Peptides 100 U 11EPSIS100 IC11EPSIS100 Each Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

Proteins, Enzymes and Peptides 3×100 U 11EPSIS103 IC11EPSIS103 Pack of 3 Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Storage and Handling Store at –20°C.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty